Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185022.1 | NP_001171951.1 | 3 | 572 | ||
NM_001185023.1 | NP_001171952.1 | 2 | 1139 | |||
NM_001307.5 | NP_001298.3 | 2 | 1139 | |||
GenBank mRNA | AF093823.1 | - | 2 | 201 | ||
AJ011497.1 | - | 3 | 571 | |||
AK313958.1 | - | 2 | 667 | |||
BC001055.2 | - | 2 | 663 | 3504311 | ||
BC071844.1 | - | 2 | 477 | 6729592 | ||
BT006829.1 | - | 2 | 237 |