Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000759.3 | NP_000750.1 | 4, 5 | 564 | ||
NM_001178147.1 | NP_001171618.1 | 3, 4 | 447 | |||
NM_172219.2 | NP_757373.1 | 4, 5 | 555 | |||
NM_172220.2 | NP_757374.2 | 3, 4 | 456 | |||
NR_033662.1 | 3, 4 | 731 | ||||
GenBank mRNA | AK290247.1 | - | 3, 4 | 715 | ||
AK298116.1 | - | 2, 3 | 607 | |||
AK313509.1 | - | 4, 5 | 480 | |||
BC033245.1 | - | 3, 4 | 625 | 5434958 | ||
CB127794.1 | - | 3, 4 | 447 | |||
CD013926.1 | - | 3, 4 | 350 | |||
CR541891.1 | - | 4, 5 | 441 | |||
HM132049.1 | - | 2, 3 | 243 | |||
KP271008.1 | - | 3, 4 | 351 | |||
M17706.1 | - | 4, 5 | 483 | |||
X03438.1 | - | 4, 5 | 481 | |||
X03655.1 | - | 4, 5 | 471 |