Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000759.3 | NP_000750.1 | 5 | 703 | ||
NM_001178147.1 | NP_001171618.1 | 4 | 586 | |||
NM_172219.2 | NP_757373.1 | 5 | 694 | |||
NM_172220.2 | NP_757374.2 | 4 | 595 | |||
NR_033662.1 | 4 | 870 | ||||
GenBank mRNA | AK290247.1 | - | 4 | 854 | ||
AK298116.1 | - | 3 | 746 | |||
AK313509.1 | - | 5 | 619 | |||
BC033245.1 | - | 4 | 764 | 5434958 | ||
CD013926.1 | - | 4 | 489 | |||
CR541891.1 | - | 5 | 580 | |||
HM132049.1 | - | 3 | 382 | |||
KP271008.1 | - | 4 | 490 | |||
M17706.1 | - | 5 | 622 | |||
X03438.1 | - | 5 | 620 | |||
X03655.1 | - | 5 | 610 |