Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DFFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282669.1 | NP_001269598.1 | 6 | 1104 | ||
NM_001320132.1 | NP_001307061.1 | 6 | 1201 | |||
NM_001320136.1 | NP_001307065.1 | 6 | 1082 | |||
NM_004402.3 | NP_004393.1 | 6 | 1032 | |||
NR_104222.1 | 7 | 1169 | ||||
NR_135150.1 | 6 | 1032 | ||||
NR_135151.1 | 7 | 1119 | ||||
NR_135152.1 | 6 | 1054 | ||||
XM_017000498.1 | XP_016855987.1 | 5 | 967 | |||
GenBank mRNA | AB013918.1 | - | 6 | 695 | ||
AB028911.1 | - | 6 | 826 | |||
AB028912.1 | - | 6 | 995 | |||
AB028913.1 | - | 7 | 913 | |||
AF039210.1 | - | 6 | 695 | |||
AF064019.1 | - | 6 | 826 | |||
AF409060.1 | - | 8 | 706 | |||
AF409061.1 | - | 6 | 721 | |||
AF409062.1 | - | 6 | 749 | |||
AK290877.1 | - | 6 | 823 | |||
AK303065.1 | - | 6 | 911 | |||
AY438267.1 | - | 6 | 624 | |||
BC032827.1 | - | 7 | 981 | |||
BC048797.1 | - | 6 | 1018 | 5204996 |