Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DFFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282669.1 | NP_001269598.1 | 7 | 1260 | ||
NM_001320132.1 | NP_001307061.1 | 7 | 1357 | |||
NM_001320136.1 | NP_001307065.1 | 7 | 1238 | |||
NM_004402.3 | NP_004393.1 | 7 | 1188 | |||
NR_104222.1 | 8 | 1325 | ||||
NR_135150.1 | 8 | 1312 | ||||
NR_135151.1 | 8 | 1275 | ||||
NR_135152.1 | 7 | 1210 | ||||
XM_017000498.1 | XP_016855987.1 | 6 | 1123 | |||
XM_017000499.1 | XP_016855988.1 | 6 | 1139 | |||
XM_017000500.1 | XP_016855989.1 | 6 | 894 | |||
GenBank mRNA | AB013918.1 | - | 7 | 851 | ||
AB028911.1 | - | 8 | 1105 | |||
AB028912.1 | - | 7 | 1151 | |||
AB028913.1 | - | 8 | 1069 | |||
AF039210.1 | - | 7 | 851 | |||
AF064019.1 | - | 7 | 982 | |||
AF409060.1 | - | 9 | 862 | |||
AF409061.1 | - | 7 | 877 | |||
AF409062.1 | - | 7 | 905 | |||
AK290877.1 | - | 7 | 979 | |||
AK303065.1 | - | 7 | 1067 | |||
AY438267.1 | - | 7 | 780 | |||
AY438268.1 | - | 4 | 403 | |||
BC032827.1 | - | 8 | 1137 | |||
BC048797.1 | - | 7 | 1174 | 5204996 |