Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYB5R3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000398.6 | NP_000389.1 | 4 | 353 | ||
NM_001129819.2 | NP_001123291.1 | 4 | 597 | |||
NM_001171660.1 | NP_001165131.1 | 4 | 493 | |||
NM_001171661.1 | NP_001165132.1 | 5 | 413 | |||
NM_007326.4 | NP_015565.1 | 4 | 352 | |||
GenBank mRNA | AF361370.1 | - | 4 | 288 | ||
AJ010116.1 | - | 4 | 266 | |||
AJ010117.1 | - | 4 | 266 | |||
AJ010118.1 | - | 4 | 266 | |||
AK124528.1 | - | 1 | 2403 | |||
AK302204.1 | - | 4 | 492 | |||
BC004821.1 | - | 4 | 297 | 3450773 | ||
BM564146.1 | - | 5 | 403 | |||
BT009821.1 | - | 4 | 269 | |||
BX334220.2 | - | 4 | 354 | |||
BX366240.2 | - | 4 | 353 | |||
CB118906.1 | - | 4 | 341 | |||
CR456435.1 | - | 4 | 275 | |||
DA268749.1 | - | 4 | 287 | |||
DA966919.1 | - | 4 | 287 | |||
DB029897.1 | - | 4 | 352 | |||
M16461.1 | - | 3 | 250 | |||
M16462.1 | - | 3 | 188 | |||
Y09501.1 | - | 4 | 288 |