Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052838.4 | NP_443070.5 | 8, 9 | 846 | ||
XM_017022991.1 | XP_016878480.1 | 8, 9 | 826 | |||
GenBank mRNA | AF085235.1 | - | 7, 8 | 677 | ||
AL833004.1 | - | 2, 3 | 1612 | |||
AY034176.1 | - | 7, 8 | 680 | |||
BC012161.1 | - | 6, 7 | 668 | 4576220 |