Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004405.3 | NP_004396.1 | 1 | 533 | ||
GenBank mRNA | AB208823.1 | - | 1 | 389 | ||
AK291367.1 | - | 1 | 683 | |||
AK297503.1 | - | 1 | 415 | |||
AK316129.1 | - | 1 | 683 | |||
BC032558.1 | - | 1 | 539 | 5562689 | ||
BE905163.1 | - | 1 | 533 | |||
KJ534825.1 | - | 1 | 321 | |||
L07919.1 | - | 1 | 70 |