Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001934.3 | NP_001925.2 | 1 | 408 | ||
NM_138281.2 | NP_612138.1 | 2 | 655 | |||
XM_017024291.1 | XP_016879780.1 | 3 | 701 | |||
GenBank mRNA | AF254115.1 | - | 2 | 668 | ||
BC005812.1 | - | 1 | 239 | |||
BC014419.1 | - | 1 | 230 | |||
BC016145.1 | - | 2 | 656 | 3907376 | ||
BG479840.1 | - | 1 | 231 | |||
BT006978.1 | - | 2 | 384 | |||
U31762.1 | - | 2 | 708 | |||
U73328.1 | - | 1 | 414 |