Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206927.1 | NP_001193856.1 | 67 | 9995 | ||
XM_011514318.2 | XP_011512620.1 | 66 | 9943 | |||
XM_011514319.2 | XP_011512621.1 | 66 | 9898 | |||
XM_011514320.2 | XP_011512622.1 | 65 | 9769 | |||
XM_011514321.1 | XP_011512623.1 | 64 | 9353 | |||
XM_017010325.1 | XP_016865814.1 | 67 | 10006 | |||
XM_017010326.1 | XP_016865815.1 | 67 | 10006 | |||
GenBank mRNA | AF356519.1 | - | 65 | 9489 |