Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNASE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005223.3 | NP_005214.2 | 2 | 2176 | ||
XM_005255148.3 | XP_005255205.1 | 5 | 1087 | |||
XM_006720854.3 | XP_006720917.1 | 5 | 1088 | |||
XM_011522393.2 | XP_011520695.1 | 5 | 1086 | |||
XM_017022992.1 | XP_016878481.1 | 5 | 1088 | |||
XM_017022993.1 | XP_016878482.1 | 6 | 1693 | |||
XM_017022994.1 | XP_016878483.1 | 7 | 1859 | |||
XM_017022995.1 | XP_016878484.1 | 5 | 893 | |||
XM_017022996.1 | XP_016878485.1 | 5 | 839 | |||
XM_017022997.1 | XP_016878486.1 | 6 | 1507 | |||
XM_017022998.1 | XP_016878487.1 | 4 | 1792 | |||
XM_017022999.1 | XP_016878488.1 | 3 | 830 | |||
XM_017023000.1 | XP_016878489.1 | 3 | 830 | |||
XM_017023001.1 | XP_016878490.1 | 5 | 1090 | |||
XM_017023002.1 | XP_016878491.1 | 2 | 995 | |||
XM_017023003.1 | XP_016878492.1 | 2 | 995 | |||
XM_017023004.1 | XP_016878493.1 | 5 | 1092 | |||
XM_017023005.1 | XP_016878494.1 | 6 | 1697 | |||
XM_017023007.1 | XP_016878496.1 | 5 | 1087 | |||
GenBank mRNA | AB188151.1 | - | 3 | 255 | ||
AB188152.1 | - | 2 | 309 | |||
AJ298844.1 | - | 2 | 1131 | |||
AK300914.1 | - | 2 | 280 | |||
BC029437.1 | - | 2 | 197 | 4594247 | ||
BX648257.1 | - | 2 | 262 | |||
M55983.1 | - | 2 | 228 |