Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007026.1 | NP_001007027.1 | 9 | 3681 | ||
NM_001940.3 | NP_001931.2 | 9 | 3674 | |||
GenBank mRNA | BC051795.2 | - | 9 | 3695 | 4181592 | |
BQ448220.1 | - | 2 | 683 | |||
D31840.1 | - | 9 | 3667 | |||
D38529.1 | - | 8 | 3588 | |||
U23851.1 | - | 8 | 3499 |