Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285389.2 | NP_001272318.1 | 3 | 353 | ||
NM_001285391.2 | NP_001272320.2 | 3 | 676 | |||
NM_001285392.2 | NP_001272321.1 | 3 | 470 | |||
NM_001285393.2 | NP_001272322.1 | 3 | 407 | |||
NM_004414.6 | NP_004405.3 | 3 | 679 | |||
NM_203417.2 | NP_981962.1 | 3 | 465 | |||
NM_203418.2 | NP_981963.1 | 3 | 644 | |||
XM_005260929.1 | XP_005260986.1 | 3 | 600 | |||
XM_005260930.1 | XP_005260987.1 | 3 | 588 | |||
XM_017028282.1 | XP_016883771.1 | 4 | 1005 | |||
XM_017028283.1 | XP_016883772.1 | 4 | 1005 | |||
GenBank mRNA | AF400429.1 | - | 3 | 337 | ||
AK055845.1 | - | 2 | 876 | |||
AK092184.1 | - | 3 | 600 | |||
AK131569.1 | - | 3 | 597 | |||
AK293293.1 | - | 3 | 458 | |||
AK299444.1 | - | 3 | 394 | |||
AL049369.1 | - | 1 | 1581 | |||
AL556803.3 | - | 3 | 679 | |||
AY325903.1 | - | 3 | 671 | |||
BC002864.2 | - | 3 | 439 | 3944959 | ||
BT007363.1 | - | 3 | 383 | |||
BX419260.2 | - | 3 | 597 | |||
CR456878.1 | - | 2 | 305 | |||
CR542065.1 | - | 3 | 383 | |||
DA586027.1 | - | 3 | 407 | |||
DB023862.1 | - | 3 | 470 | |||
U28833.2 | - | 3 | 353 | |||
U85265.2 | - | 3 | 464 | |||
U85266.2 | - | 3 | 407 | |||
U85267.2 | - | 3 | 526 |