Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSC22D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015881.1 | NP_001015881.1 | 2 | 188 | ||
NM_001318468.1 | NP_001305397.1 | 3 | 656 | |||
NM_001318470.1 | NP_001305399.1 | 3 | 642 | |||
NM_004089.3 | NP_004080.2 | 2 | 393 | |||
NM_198057.2 | NP_932174.1 | 2 | 709 | |||
XM_005262099.1 | XP_005262156.1 | 4 | 670 | |||
XM_005262100.1 | XP_005262157.1 | 3 | 545 | |||
XM_005262102.1 | XP_005262159.1 | 3 | 538 | |||
XM_005262103.3 | XP_005262160.1 | 3 | 648 | |||
XM_011530884.1 | XP_011529186.1 | 3 | 744 | |||
XM_017029335.1 | XP_016884824.1 | 3 | 480 | |||
GenBank mRNA | AB025432.1 | - | 2 | 376 | ||
AF153603.1 | - | 2 | 371 | |||
AF183393.1 | - | 2 | 379 | |||
AF228339.1 | - | 2 | 383 | |||
AK092645.1 | - | 3 | 595 | |||
AK092669.1 | - | 2 | 681 | |||
AK127938.1 | - | 2 | 333 | |||
AK300535.1 | - | 3 | 520 | |||
AL110191.1 | - | 2 | 392 | |||
AY007119.1 | - | 2 | 292 | |||
BC018148.1 | - | 2 | 75 | |||
BC072446.1 | - | 2 | 387 | 6165765 | ||
BM047061.1 | - | 2 | 708 | |||
BQ919216.1 | - | 188 | ||||
BX647854.1 | - | 3 | 642 | |||
CR533450.1 | - | 2 | 131 | |||
CR933650.1 | - | 2 | 395 | |||
Z50781.1 | - | 2 | 107 |