Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF1AX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001412.3 | NP_001403.1 | 7 | 4084 | ||
GenBank mRNA | AL079283.1 | - | 1 | 1116 | ||
AL832930.1 | - | 1 | 2623 | |||
BC047573.1 | - | 1 | 1933 |