Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STOM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270526.1 | NP_001257455.1 | 2 | 203 | ||
NM_001270527.1 | NP_001257456.1 | 2 | 203 | |||
NM_004099.5 | NP_004090.4 | 2 | 203 | |||
NM_198194.2 | NP_937837.1 | 2 | 203 | |||
NR_073037.1 | 3 | 333 | ||||
GenBank mRNA | AK302977.1 | - | 2 | 129 | ||
AK304449.1 | - | 3 | 277 | |||
AK304518.1 | - | 2 | 147 | |||
AK307999.1 | - | 2 | 147 | |||
BC010703.1 | - | 2 | 125 | 3873494 | ||
BG701899.1 | - | 2 | 156 | |||
BI553227.1 | - | 2 | 132 | |||
BM451470.1 | - | 167 | ||||
BP208523.1 | - | 2 | 130 | |||
CR542100.1 | - | 2 | 86 | |||
DC368197.1 | - | 2 | 203 | |||
M81635.1 | - | 2 | 107 | |||
X60067.1 | - | 2 | 86 |