Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FABP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001443.2 | NP_001434.1 | 3, 4 | 444 | ||
GenBank mRNA | BC022287.1 | - | 4, 5 | 429 | ||
BC032801.1 | - | 3, 4 | 355 | 5190267 | ||
BP263155.1 | - | 3, 4 | 288 | |||
CR542090.1 | - | 3, 4 | 330 | |||
M10050.1 | - | 3, 4 | 372 | |||
M10617.1 | - | 3, 4 | 345 |
Gene Symbol: PRDM10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK000234.1 | - | 2790 |