Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004108.2 | NP_004099.2 | 8 | 754 | ||
NM_015837.2 | NP_056652.1 | 7 | 640 | |||
XM_006717015.3 | XP_006717078.1 | 7 | 1046 | |||
XM_011518392.2 | XP_011516694.1 | 8 | 1162 | |||
GenBank mRNA | AI052239.1 | - | 1 | 358 | ||
AK290843.1 | - | 8 | 758 | |||
AK308476.1 | - | 7 | 1223 | |||
BC069572.1 | - | 8 | 754 | 7262417 | ||
BC069825.1 | - | 8 | 754 | 7262429 | ||
D49353.1 | - | 8 | 754 | |||
JB054537.1 | - | 8 | 754 |