Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FGFR1 SIRNA Assays ›
Gene Symbol:
FGFR1
Gene Name:
fibroblast growth factor receptor 1
Gene Aliases:
BFGFR, CD331, CEK, ECCL, FGFBR, FGFR-1, FLG, FLT-2, FLT2, HBGFR, HH2, HRTFDS, KAL2, N-SAM, OGD, bFGF-R-1
Chromosome Location:
Chr.8: 38411138 - 38468834 on Build GRCh38
UniGene ID:
Hs.264887
Species:
Human
siRNA ID
HSS142012
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FGFR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001174063.1
NP_001167534.1
5
1538
NM_001174064.1
NP_001167535.1
6
1606
NM_001174065.1
NP_001167536.1
5
852
NM_001174066.1
NP_001167537.1
4
591
NM_001174067.1
NP_001167538.1
6
1012
NM_015850.3
NP_056934.2
5
1532
NM_023105.2
NP_075593.1
4
1271
NM_023106.2
NP_075594.1
4
1265
NM_023110.2
NP_075598.2
5
1538
XM_006716303.2
XP_006716366.1
5
1339
XM_006716304.1
XP_006716367.1
5
855
XM_006716306.2
XP_006716369.1
5
1333
XM_006716307.1
XP_006716370.1
5
849
XM_006716309.3
XP_006716372.1
5
1309
XM_006716310.2
XP_006716373.1
4
1072
XM_006716311.1
XP_006716374.1
4
470
XM_006716312.1
XP_006716375.1
4
540
XM_006716313.2
XP_006716376.1
4
1066
XM_006716314.1
XP_006716377.1
4
544
XM_011544443.2
XP_011542745.1
5
844
XM_011544444.1
XP_011542746.1
5
833
XM_011544445.2
XP_011542747.1
5
852
XM_011544446.2
XP_011542748.1
5
839
XM_011544447.2
XP_011542749.1
5
839
XM_011544448.1
XP_011542750.1
4
443
XM_011544449.1
XP_011542751.1
4
437
XM_011544450.2
XP_011542752.1
4
437
XM_011544451.1
XP_011542753.1
3
306
XM_011544452.2
XP_011542754.1
5
927
XM_017013219.1
XP_016868708.1
5
840
XM_017013220.1
XP_016868709.1
5
833
XM_017013221.1
XP_016868710.1
6
1048
XM_017013222.1
XP_016868711.1
5
1339
XM_017013223.1
XP_016868712.1
5
849
XM_017013224.1
XP_016868713.1
5
1333
XM_017013225.1
XP_016868714.1
5
816
XM_017013226.1
XP_016868715.1
4
443
XM_017013227.1
XP_016868716.1
4
437
XM_017013228.1
XP_016868717.1
4
1066
XM_017013230.1
XP_016868719.1
3
397
XM_017013231.1
XP_016868720.1
5
937
GenBank mRNA
AB208919.1
-
6
1393
AK222718.1
-
5
1331
AK223126.1
-
4
1066
AK291754.1
-
5
731
AK292470.1
-
4
591
AK309947.1
-
6
891
BC015035.1
-
6
1266
3911101
BC018128.1
-
5
1296
3896359
BC091494.1
-
3
402
30387634
BX649167.1
-
1
2880
EU826596.1
-
3
323
FJ809916.1
-
5
1541
FJ809917.1
-
5
1541
KU178066.1
-
4
596
KU178067.1
-
3
329
KU178068.1
-
3
323
KU178069.1
-
3
329
KU178070.1
-
3
323
M34185.1
-
4
584
M34186.1
-
4
849
M34187.1
-
4
745
M34188.1
-
4
845
M34641.1
-
4
599
M37722.1
-
4
551
M60485.1
-
6
1261
M63887.1
-
4
596
M63888.1
-
4
596
M63889.1
-
4
473
X51803.1
-
5
713
X52833.1
-
4
683
X57118.1
-
5
734
X57119.1
-
4
491
X57120.1
-
5
752
X57121.1
-
5
758
X57122.1
-
4
485
X66945.1
-
5
1322
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
MAPK cascade
skeletal system development
angiogenesis
ureteric bud development
in utero embryonic development
organ induction
neuron migration
positive regulation of mesenchymal cell proliferation
chondrocyte differentiation
transcription, DNA-templated
protein phosphorylation
sensory perception of sound
positive regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
positive regulation of phospholipase activity
positive regulation of phospholipase C activity
positive regulation of neuron projection development
regulation of phosphatidylinositol 3-kinase signaling
positive regulation of phosphatidylinositol 3-kinase signaling
cell migration
peptidyl-tyrosine phosphorylation
stem cell population maintenance
motogenic signaling involved in postnatal olfactory bulb interneuron migration
ventricular zone neuroblast division
embryonic limb morphogenesis
midbrain development
neuron projection development
fibroblast growth factor receptor signaling pathway involved in orbitofrontal cortex development
phosphatidylinositol-3-phosphate biosynthetic process
inner ear morphogenesis
outer ear morphogenesis
middle ear morphogenesis
chordate embryonic development
positive regulation of MAP kinase activity
positive regulation of MAPK cascade
positive regulation of GTPase activity
regulation of cell differentiation
positive regulation of neuron differentiation
negative regulation of osteoblast differentiation
positive regulation of cell cycle
protein autophosphorylation
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
paraxial mesoderm development
regulation of lateral mesodermal cell fate specification
cell maturation
skeletal system morphogenesis
mesenchymal cell differentiation
regulation of sensory perception of pain
positive regulation of cardiac muscle cell proliferation
auditory receptor cell development
branching involved in salivary gland morphogenesis
lung-associated mesenchyme development
regulation of branching involved in salivary gland morphogenesis by mesenchymal-epithelial signaling
regulation of stem cell proliferation
positive regulation of MAPKKK cascade by fibroblast growth factor receptor signaling pathway
positive regulation of endothelial cell chemotaxis to fibroblast growth factor
regulation of extrinsic apoptotic signaling pathway in absence of ligand
glycoprotein binding
protein tyrosine kinase activity
fibroblast growth factor-activated receptor activity
Ras guanyl-nucleotide exchange factor activity
protein binding
ATP binding
heparin binding
1-phosphatidylinositol-3-kinase activity
fibroblast growth factor binding
protein complex binding
identical protein binding
protein homodimerization activity
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
cell adhesion molecule binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-8tsm6:80/100.66.77.47:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline