Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004467.3 | NP_004458.3 | 2 | 153 | ||
NM_147203.2 | NP_671736.2 | 2 | 241 | |||
NM_201552.1 | NP_963846.1 | 3 | 280 | |||
NM_201553.1 | NP_963847.1 | 3 | 368 | |||
XM_017013232.1 | XP_016868721.1 | 3 | 1130 | |||
GenBank mRNA | AF042354.1 | - | 2 | 188 | ||
BG565379.1 | - | 3 | 306 | |||
BG566131.1 | - | 3 | 263 | |||
BG567956.1 | - | 2 | 242 | |||
BG618053.1 | - | 2 | 154 |