Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004467.3 | NP_004458.3 | 7, 8 | 889 | ||
NM_147203.2 | NP_671736.2 | 7, 8 | 977 | |||
NM_201552.1 | NP_963846.1 | 8, 9 | 1016 | |||
NM_201553.1 | NP_963847.1 | 8, 9 | 1104 | |||
XM_017013232.1 | XP_016868721.1 | 8, 9 | 1866 | |||
XM_017013233.1 | XP_016868722.1 | 6, 7 | 941 | |||
GenBank mRNA | AF042354.1 | - | 7, 8 | 925 | ||
AF168954.1 | - | 7, 8 | 912 | |||
AK225616.1 | - | 7, 8 | 875 | |||
AK225619.1 | - | 7, 8 | 864 | |||
AK289448.1 | - | 7, 8 | 875 | |||
AX772838.1 | - | 8, 9 | 969 | |||
BC007047.1 | - | 7, 8 | 885 | 4071810 | ||
BT006635.1 | - | 6, 7 | 780 | |||
D14446.1 | - | 7, 8 | 858 | |||
D87342.1 | - | 6, 7 | 785 |