Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001452.1 | NP_001443.1 | 1 | 732 | ||
GenBank mRNA | U13220.1 | - | 1 | 732 |