Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001453.2 | NP_001444.2 | 1 | 1629 | ||
GenBank mRNA | BC134421.1 | - | 2 | 717 |