Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005251.2 | NP_005242.1 | 1 | 145 | ||
GenBank mRNA | BC113437.1 | - | 1 | 145 | 8322489 | |
BC113439.1 | - | 1 | 145 | 8322491 |