Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000149.3 | NP_000140.1 | 3 | 829 | ||
NM_001097639.1 | NP_001091108.1 | 3 | 481 | |||
NM_001097640.1 | NP_001091109.1 | 3 | 427 | |||
NM_001097641.1 | NP_001091110.1 | 2 | 288 | |||
GenBank mRNA | AF131913.1 | - | 2 | 360 | ||
AK291852.1 | - | 3 | 456 | |||
AK313575.1 | - | 3 | 478 | |||
BC074836.2 | - | 2 | 233 | 30915328 | ||
BC074837.2 | - | 2 | 233 | 30915548 | ||
BC108675.1 | - | 2 | 285 | 5187745 | ||
DA876997.1 | - | 3 | 402 | |||
U27326.1 | - | 3 | 420 | |||
U27327.1 | - | 3 | 474 | |||
U27328.1 | - | 4 | 823 | |||
X53578.1 | - | 2 | 263 |