Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFI6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002038.3 | NP_002029.3 | 3 | 243 | ||
NM_022872.2 | NP_075010.1 | 3 | 255 | |||
NM_022873.2 | NP_075011.1 | 3 | 267 | |||
GenBank mRNA | AK024814.1 | - | 3 | 223 | ||
AK314488.1 | - | 3 | 234 | |||
BC011601.2 | - | 3 | 194 | 3140035 | ||
BC015603.2 | - | 3 | 196 | 4654295 | ||
BT006850.1 | - | 2 | 120 | |||
X02492.1 | - | 3 | 189 |