Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFI6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002038.3 | NP_002029.3 | 5 | 440 | ||
NM_022872.2 | NP_075010.1 | 5 | 452 | |||
NM_022873.2 | NP_075011.1 | 5 | 464 | |||
GenBank mRNA | AK024814.1 | - | 5 | 420 | ||
AK314488.1 | - | 5 | 431 | |||
BC011601.2 | - | 5 | 391 | 3140035 | ||
BC015603.2 | - | 5 | 393 | 4654295 | ||
BT006850.1 | - | 4 | 317 | |||
X02492.1 | - | 5 | 386 |