Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000403.3 | NP_000394.2 | 6 | 690 | ||
NM_001008216.1 | NP_001008217.1 | 6 | 625 | |||
NM_001127621.1 | NP_001121093.1 | 5 | 666 | |||
GenBank mRNA | AK057302.1 | - | 4 | 861 | ||
AK314397.1 | - | 5 | 638 | |||
BC001273.1 | - | 5 | 546 | 3459004 | ||
BC050685.2 | - | 6 | 549 | 6161561 | ||
DQ233668.1 | - | 4 | 456 | |||
L41668.1 | - | 6 | 549 |