Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GANC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301409.1 | NP_001288338.1 | 6, 7 | 1209 | ||
NM_198141.2 | NP_937784.2 | 5, 6 | 822 | |||
GenBank mRNA | AF525397.1 | - | 1, 2 | 75 | ||
AF545045.1 | - | 5, 6 | 516 | |||
AF545046.1 | - | 5, 6 | 516 | |||
AK074037.1 | - | 5, 6 | 714 | |||
AK074330.1 | - | 6, 7 | 1211 | |||
BC059406.1 | - | 5, 6 | 694 | 30330796 | ||
BC093833.1 | - | 5, 6 | 539 | 7939678 | ||
BC112051.1 | - | 5, 6 | 539 | 8327519 |