Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286205.1 | NP_001273134.1 | 7 | 1127 | ||
NM_001286208.1 | NP_001273137.1 | 6 | 1072 | |||
NM_001286209.1 | NP_001273138.1 | 7 | 1019 | |||
NM_001481.2 | NP_001472.1 | 7 | 1004 | |||
XM_005256304.4 | XP_005256361.1 | 8 | 1659 | |||
XM_005256309.4 | XP_005256366.1 | 9 | 1576 | |||
XM_006721175.3 | XP_006721238.1 | 7 | 1057 | |||
XM_011522990.2 | XP_011521292.1 | 7 | 1078 | |||
XM_011522991.1 | XP_011521293.1 | 8 | 1233 | |||
XM_011522992.2 | XP_011521294.1 | 6 | 1026 | |||
XM_017023122.1 | XP_016878611.1 | 9 | 1384 | |||
XM_017023123.1 | XP_016878612.1 | 7 | 1083 | |||
XM_017023124.1 | XP_016878613.1 | 6 | 870 | |||
XM_017023125.1 | XP_016878614.1 | 6 | 1002 | |||
GenBank mRNA | AF050079.1 | - | 7 | 1003 | ||
AK293958.1 | - | 7 | 1034 | |||
AK294050.1 | - | 6 | 853 | |||
AK297857.1 | - | 7 | 987 | |||
AK315261.1 | - | 7 | 952 | |||
BC036781.1 | - | 5 | 2082 | |||
BC104785.1 | - | 7 | 989 | 8143788 | ||
BC112121.1 | - | 7 | 989 | 8327589 | ||
BX647947.1 | - | 6 | 990 | |||
CR749499.1 | - | 5 | 1682 |