Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNG11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004126.3 | NP_004117.1 | 1 | 446 | ||
GenBank mRNA | AF493878.1 | - | 1 | 12 | ||
AK312037.1 | - | 1 | 96 | |||
BC009709.1 | - | 1 | 363 | 3879341 | ||
BG255463.1 | - | 455 | ||||
BT007141.1 | - | 1 | 12 | |||
U31384.1 | - | 1 | 119 |