Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012009.1 | XP_016867498.1 | 2, 3 | 238 | ||
GenBank mRNA | AF493869.1 | - | 1, 2 | 103 | ||
BC029367.1 | - | 2, 3 | 251 | 4604788 | ||
BG435040.1 | - | 2, 3 | 251 | |||
BQ637754.1 | - | 2, 3 | 150 | |||
CR456912.1 | - | 1, 2 | 103 | |||
EL950965.1 | - | 2, 3 | 178 | |||
S62027.1 | - | 1, 2 | 111 |