Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006142.3 | NP_006133.1 | 1 | 212 | ||
GenBank mRNA | AF029082.1 | - | 1 | 212 | ||
BC000329.2 | - | 1 | 192 | 2822397 | ||
BC000995.2 | - | 1 | 189 | 3445915 | ||
BC001550.1 | - | 1 | 194 | 3457665 | ||
BC002995.1 | - | 1 | 192 | 2822397 | ||
BC023552.2 | - | 1 | 216 | 3534328 | ||
CR541905.1 | - | 1 | 141 | |||
CR541926.1 | - | 1 | 141 | |||
M93010.1 | - | 1 | 151 |