Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000174.4 | NP_000165.1 | 3 | 740 | ||
XM_005247374.3 | XP_005247431.1 | 5 | 1036 | |||
XM_011512701.1 | XP_011511003.1 | 6 | 1118 | |||
XM_011512702.1 | XP_011511004.1 | 5 | 993 | |||
GenBank mRNA | AA701315.1 | - | 1 | 150 | ||
BC030229.1 | - | 3 | 705 | 5226377 | ||
BM971739.1 | - | 1 | 168 | |||
M25827.1 | - | 1 | 482 | |||
X52997.1 | - | 3 | 740 |