Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002083.3 | NP_002074.2 | 1 | 242 | ||
NR_046320.1 | 1 | 242 | ||||
NR_046321.1 | 1 | 242 | ||||
GenBank mRNA | AK225871.1 | - | 1 | 82 | ||
BC005277.1 | - | 1 | 82 | 3996732 | ||
BC016756.1 | - | 1 | 96 | 3681457 | ||
BC022820.1 | - | 1 | 65 | 5429926 | ||
BC067221.1 | - | 1 | 86 | 6668339 | ||
BF207526.1 | - | 82 | ||||
BG429974.1 | - | 1 | 243 | |||
X53463.1 | - | 1 | 114 | |||
X68314.1 | - | 1 | 97 |