Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039847.2 | NP_001034936.1 | 2 | 280 | ||
NM_001039848.2 | NP_001034937.1 | 2 | 358 | |||
NM_002085.4 | NP_002076.2 | 2 | 280 | |||
GenBank mRNA | AK225859.1 | - | 2 | 199 | ||
AL558552.3 | - | 2 | 133 | |||
BC010157.2 | - | 2 | 108 | |||
BC011836.2 | - | 2 | 201 | 3945177 | ||
BC021567.1 | - | 2 | 194 | 4581069 | ||
BC022071.1 | - | 2 | 217 | 4821595 | ||
BC032695.1 | - | 2 | 201 | 5227887 | ||
BC039849.1 | - | 2 | 269 | 5744771 | ||
BC046163.1 | - | 2 | 176 | 5742889 | ||
BC058329.1 | - | 2 | 197 | |||
BI561548.1 | - | 2 | 276 | |||
BM764133.1 | - | 2 | 359 | |||
BQ648717.1 | - | 2 | 145 | |||
CF127510.1 | - | 2 | 292 | |||
HY049693.1 | - | 2 | 281 | |||
X71973.1 | - | 2 | 242 |