Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRINA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000837.1 | NP_000828.1 | 5 | 1015 | ||
NM_001009184.1 | NP_001009184.1 | 5 | 903 | |||
GenBank mRNA | AB097026.1 | - | 5 | 827 | ||
AK127640.1 | - | 5 | 1051 | |||
AK294127.1 | - | 6 | 760 | |||
BC041788.1 | - | 5 | 861 | 4738372 | ||
BC065834.1 | - | 4 | 769 | |||
BC084553.1 | - | 4 | 752 | 6206624 | ||
U44954.1 | - | 2 | 86 |