Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001511.3 | NP_001502.1 | 4 | 623 | ||
NR_046035.1 | 4 | 646 | ||||
GenBank mRNA | BC011976.1 | - | 4 | 611 | 3856841 | |
BM554448.1 | - | 4 | 636 | |||
BQ000742.1 | - | 1 | 493 | |||
J03561.1 | - | 4 | 597 | |||
X12510.1 | - | 4 | 561 |