Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDIA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005313.4 | NP_005304.3 | 13 | 2203 | ||
GenBank mRNA | BM854408.1 | - | 1 | 265 |