Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000846.4 | NP_000837.3 | 6 | 661 | ||
XM_011514532.2 | XP_011512834.1 | 6 | 653 | |||
GenBank mRNA | AK313601.1 | - | 6 | 573 | ||
BC002895.2 | - | 6 | 624 | 3943417 | ||
BE795593.1 | - | 6 | 604 | |||
BE796134.1 | - | 6 | 605 | |||
M16594.1 | - | 6 | 577 |