Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001312660.1 | NP_001299589.1 | 8, 9 | 943 | ||
NM_145870.2 | NP_665877.1 | 7, 8 | 757 | |||
NM_145871.2 | NP_665878.2 | 6, 7 | 631 | |||
XM_011536670.2 | XP_011534972.1 | 7, 8 | 1358 | |||
XM_011536671.2 | XP_011534973.1 | 6, 7 | 541 | |||
XM_017021243.1 | XP_016876732.1 | 6, 7 | 1028 | |||
GenBank mRNA | AB209360.1 | - | 4, 5 | 4883 | ||
AJ001838.1 | - | 7, 8 | 578 | |||
AK315154.1 | - | 8, 9 | 834 | |||
AK315609.1 | - | 7, 8 | 601 | |||
BC001453.1 | - | 7, 8 | 633 | 3139094 | ||
BE958109.1 | - | 438 | ||||
BG330365.1 | - | 6, 7 | 493 | |||
CR456987.1 | - | 6, 7 | 475 | |||
U86529.1 | - | 7, 8 | 578 |