Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001312660.1 | NP_001299589.1 | 10 | 1041 | ||
NM_145870.2 | NP_665877.1 | 9 | 855 | |||
NM_145871.2 | NP_665878.2 | 8 | 729 | |||
XM_011536670.2 | XP_011534972.1 | 9 | 1456 | |||
XM_011536671.2 | XP_011534973.1 | 8 | 639 | |||
XM_017021243.1 | XP_016876732.1 | 8 | 1126 | |||
GenBank mRNA | AB209360.1 | - | 6 | 4981 | ||
AJ001838.1 | - | 9 | 676 | |||
AK094344.1 | - | 2 | 1969 | |||
AK315154.1 | - | 10 | 932 | |||
AK315609.1 | - | 9 | 699 | |||
BC001453.1 | - | 9 | 731 | 3139094 | ||
BE958109.1 | - | 537 | ||||
BG330365.1 | - | 8 | 590 | |||
CR456987.1 | - | 8 | 573 | |||
U86529.1 | - | 9 | 676 |