Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000858.5 | NP_000849.1 | 8 | 800 | ||
NM_001159390.1 | NP_001152862.1 | 7 | 635 | |||
NM_001159391.1 | NP_001152863.1 | 7 | 549 | |||
NM_001242839.1 | NP_001229768.1 | 7 | 575 | |||
XM_005273104.4 | XP_005273161.1 | 9 | 969 | |||
XM_017001092.1 | XP_016856581.1 | 8 | 1288 | |||
XM_017001095.1 | XP_016856584.1 | 8 | 745 | |||
XM_017001096.1 | XP_016856585.1 | 7 | 585 | |||
GenBank mRNA | AK124677.1 | - | 3347 | |||
AK303845.1 | - | 7 | 588 | |||
AW083682.1 | - | 2 | 352 | |||
BC006249.2 | - | 8 | 738 | 3946293 | ||
BC007369.2 | - | 6 | 452 | |||
BC009914.2 | - | 8 | 712 | 2821563 | ||
BC091480.1 | - | 7 | 555 | 30340102 | ||
BE780558.1 | - | 7 | 522 | |||
BI771824.1 | - | 7 | 575 | |||
CR456836.1 | - | 6 | 497 | |||
CR542006.1 | - | 6 | 497 | |||
L76200.1 | - | 6 | 497 | |||
U66895.1 | - | 6 | 503 |