Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GYPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256584.1 | NP_001243513.1 | 5 | 1518 | ||
NM_002101.4 | NP_002092.1 | 4 | 700 | |||
NM_016815.3 | NP_058131.1 | 3 | 643 | |||
XM_006712460.3 | XP_006712523.1 | 4 | 568 | |||
GenBank mRNA | AK312032.1 | - | 4 | 488 | ||
BC104246.1 | - | 4 | 501 | 40033899 | ||
BC104247.1 | - | 4 | 501 | 40033903 | ||
BC106051.1 | - | 4 | 532 | 4707035 | ||
CB992018.1 | - | 3 | 428 | |||
M11802.1 | - | 4 | 371 | |||
M28335.1 | - | 4 | 365 | |||
M36284.1 | - | 4 | 371 | |||
X12496.1 | - | 4 | 491 | |||
X51973.1 | - | 3 | 314 |