Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GYPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002102.3 | NP_002093.2 | 3 | 247 | ||
NM_198682.2 | NP_941391.2 | 3 | 247 | |||
XM_017008139.1 | XP_016863628.1 | 4 | 4174 | |||
XM_017008140.1 | XP_016863629.1 | 4 | 4174 | |||
GenBank mRNA | BC017864.1 | - | 3 | 243 | 4686481 | |
BC064396.1 | - | 3 | 204 | |||
BG528203.1 | - | 3 | 243 | |||
BX091225.1 | - | 3 | 215 | |||
M29610.1 | - | 3 | 247 | |||
N38976.1 | - | 2 | 250 |