Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2AFZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002106.3 | NP_002097.1 | 5 | 797 | ||
GenBank mRNA | AK056803.1 | - | 2 | 1692 | ||
BC018002.1 | - | 5 | 717 | 4271353 | ||
BC020936.1 | - | 5 | 717 | 4281642 | ||
BC103743.1 | - | 5 | 832 | 6175124 | ||
M37583.1 | - | 5 | 718 | |||
X52317.1 | - | 5 | 718 |