Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MNX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005515.3 | NP_005506.3 | 1 | 858 | ||
GenBank mRNA | AF107457.1 | - | 1 | 742 | ||
X56537.1 | - | 1 | 65 |