Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLA-DRB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001298.1 | 6 | 970 | |||
GenBank mRNA | AF005487.1 | - | 6 | 970 | ||
L76565.1 | - | 6 | 1041 |
Gene Symbol: HLA-DRB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021983.4 | NP_068818.4 | 6 | 940 | ||
GenBank mRNA | AF048707.1 | - | 5 | 730 | ||
AI829825.1 | - | 1 | 250 | |||
AK292151.1 | - | 7 | 920 | |||
BC005312.1 | - | 7 | 932 | 3933019 | ||
M15178.1 | - | 7 | 811 | |||
M16942.1 | - | 7 | 897 |