Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319077.1 | NP_001306006.1 | 5 | 523 | ||
NM_001319078.1 | NP_001306007.1 | 4 | 444 | |||
NM_001319079.1 | NP_001306008.1 | 5 | 556 | |||
NM_001319080.1 | NP_001306009.1 | 5 | 555 | |||
NM_001319081.1 | NP_001306010.1 | 4 | 442 | |||
NM_001319082.1 | NP_001306011.1 | 5 | 578 | |||
NM_002131.3 | NP_002122.1 | 5 | 525 | |||
NM_145899.2 | NP_665906.1 | 5 | 558 | |||
NM_145901.2 | NP_665908.1 | 4 | 725 | |||
NM_145902.2 | NP_665909.1 | 4 | 692 | |||
NM_145903.2 | NP_665910.1 | 4 | 411 | |||
NM_145905.2 | NP_665912.1 | 4 | 414 | |||
GenBank mRNA | AK096863.1 | - | 6 | 464 | ||
AK308245.1 | - | 4 | 698 | |||
BC004924.2 | - | 5 | 405 | 3605116 | ||
BC008832.2 | - | 5 | 438 | 4308914 | ||
BC015789.1 | - | 4 | 414 | 4866014 | ||
BC063434.1 | - | 4 | 330 | 5399570 | ||
BC067083.1 | - | 5 | 413 | 4110270 | ||
BC071864.1 | - | 5 | 433 | 6083428 | ||
BT006774.1 | - | 3 | 203 | |||
CX785762.1 | - | 4 | 346 | |||
DA467015.1 | - | 5 | 483 | |||
DA574958.1 | - | 4 | 393 | |||
DA936387.1 | - | 5 | 480 | |||
DB029421.1 | - | 5 | 505 | |||
KF731633.1 | - | 3 | 236 | |||
M23614.1 | - | 4 | 443 | |||
M23615.1 | - | 4 | 410 | |||
M23616.1 | - | 5 | 445 | |||
M23617.1 | - | 4 | 338 | |||
M23618.1 | - | 3 | 245 | |||
M23619.1 | - | 5 | 548 | |||
X14957.1 | - | 5 | 467 | |||
X14958.1 | - | 5 | 434 |