Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003810.1 | NP_001003810.1 | 2 | 580 | ||
NM_002138.3 | NP_002129.2 | 3 | 637 | |||
NM_031369.2 | NP_112737.1 | 2 | 580 | |||
NM_031370.2 | NP_112738.1 | 3 | 637 | |||
GenBank mRNA | AF039575.1 | - | 3 | 295 | ||
AK057836.1 | - | 3 | 598 | |||
AK292707.1 | - | 2 | 575 | |||
AK300149.1 | - | 3 | 469 | |||
BC002401.1 | - | 3 | 383 | 2822230 | ||
BC023977.2 | - | 3 | 612 | 2961272 | ||
BC026015.2 | - | 3 | 612 | 2961272 | ||
D55671.1 | - | 2 | 56 | |||
D55672.1 | - | 3 | 604 | |||
D55673.1 | - | 2 | 47 | |||
D55674.1 | - | 3 | 583 | |||
M94630.1 | - | 3 | 277 | |||
X03910.1 | - | 3 | 313 |